Skip to Content
Merck
Ammonium sulfate anhydrous, Redi-Dri™, ReagentPlus®, ≥99.0%

Ammonium sulfate

Ammonium sulfate anhydrous, Redi-Dri™, ReagentPlus®, ≥99.0%
Synonym(s):
Mascagnite, Ammonium sulfate
Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14

Products

Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique