Skip to Content
Merck

Skip To

1374259

USP

Magnesium citrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Magnesium citrate (3:2)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 480,00

About This Item

Empirical Formula (Hill Notation):
C12H10Mg3O14
CAS Number:
Molecular Weight:
451.11
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 480,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

magnesium citrate

packaging

pkg of 500 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Mg+2].[Mg+2].[Mg+2].[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-].[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-]

InChI

1S/2C6H8O7.3Mg/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;;3*+2/p-6

InChI key

PLSARIKBYIPYPF-UHFFFAOYSA-H

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Magnesium citrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
108697913742811133591
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

room temp

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

packaging

pkg of 500 mg

packaging

pkg of 200 mg

packaging

pkg of 500 mg

packaging

pkg of 500 mg

API family

magnesium citrate

API family

magnesium citrate

API family

magnesium oxide

API family

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters