Skip to Content
Merck

Skip To

1374281

USP

Magnesium Oxide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Magnesium oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 407,00

About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 407,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

magnesium oxide

packaging

pkg of 500 mg

manufacturer/tradename

USP

mp

2852 °C (lit.)

application(s)

pharmaceutical (small molecules)

format

neat

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Magnesium Oxide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Magnesium Oxide Capsules
  • Magnesium Oxide Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.058671.05862243388
application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical
solid formulation

application(s)

pharma/biopharma processes

application(s)

-

format

neat

format

-

format

-

format

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

packaging

pkg of 500 mg

packaging

-

packaging

-

packaging

-

mp

2852 °C (lit.)

mp

2800 °C

mp

2800 °C

mp

2852 °C (lit.)

API family

magnesium oxide

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Magnesium Oxide


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters