Skip to Content
Merck

Skip To

1372468

USP

Polyoxyl 23 lauryl ether

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Polyoxyethylene(23) lauryl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
0.5 g
Check Cart for Availability
€ 391,00

About This Item

Linear Formula:
C12H25(OCH2CH2)nOH, n~23
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 391,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

polyoxyl

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O(CCO)CCCCCCCCCCCC

InChI

1S/C14H30O2/c1-2-3-4-5-6-7-8-9-10-11-13-16-14-12-15/h15H,2-14H2,1H3

InChI key

SFNALCNOMXIBKG-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Polyoxyl 23 lauryl ether USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Polyoxyl Lauryl Ether

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P9769203728203724
grade

pharmaceutical primary standard

grade

-

grade

-

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

-

format

neat

format

-

format

-

format

-

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

15-25°C

API family

polyoxyl

API family

-

API family

-

API family

-


signalword

Warning

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Exclamation mark

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1372468-0.5G04061838669858

Questions

Reviews

No rating value

Active Filters