Skip to Content
Merck
All Photos(1)

Key Documents

203728

Millipore

BRIJ® 35 Detergent, Protein Grade®, 10% Solution, Sterile-Filtered

Non-ionic detergent useful for the isolation of functional membrane complexes.

Synonym(s):

BRIJ® 35 Detergent, Protein Grade®, 10% Solution, Sterile-Filtered, Polyoxyethyleneglycol Dodecyl Ether, Polyoxyethylene (23) Lauryl Ether

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
Inquire


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
Inquire

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21


Estimated to ship on24 May 2025


Request a Bulk Order

description

Merck USA index - 14, 7558

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

40

impurities

<200 μM Aldehyde
<200 μM peroxide

CMC

92 μM

shipped in

ambient

storage temp.

2-8°C

SMILES string

O(CCO)CCCCCCCCCCCC

InChI

1S/C14H30O2/c1-2-3-4-5-6-7-8-9-10-11-13-16-14-12-15/h15H,2-14H2,1H3

InChI key

SFNALCNOMXIBKG-UHFFFAOYSA-N

General description

Conductivity: <50 µmhos
Non-ionic detergent useful for the isolation of functional membrane complexes (aggregation number: 40; CMC: 92 mM; HLB number: 16.9). Can be used in protocols that require UV monitoring of the solubilized proteins because of its low UV absorbance (conductivity: <50 µmhos). Has been purified to reduce levels of contaminating aldehydes, metals, peroxides and salts.

Warning

Toxicity: Harmful (C)

Physical form

10% aqueous solution.

Other Notes

Lever, M. 1997. Anal. Biochem.83, 274.
Ashani, Y., and Catravas, G.N. 1980. Anal. Biochem.109, 55.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>300.2 °F

Flash Point(C)

> 149 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service