Skip to Content
Merck

Skip To

1225237

USP

Dopamine Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(3,4-Dimethoxyphenyl)ethan-1-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 1.500,00

About This Item

Empirical Formula (Hill Notation):
C10H15NO2
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 1.500,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


API family

dopamine

packaging

pkg of 100 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NCCc1cc(c(cc1)OC)OC

InChI

1S/C10H15NO2/c1-12-9-4-3-8(5-6-11)7-10(9)13-2/h3-4,7H,5-6,11H2,1-2H3

InChI key

ANOUKFYBOAKOIR-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Dopamine Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Dopamine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
122521512252261140445
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

dopamine

API family

dopamine

API family

dopamine

API family

clonidine

packaging

pkg of 100 mg

packaging

pkg of 30 mg

packaging

pkg of 30 mg

packaging

pkg of 15 mg

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

flash_point_f

257.0 °F

flash_point_c

125 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1225237-100MG04065267682631

Questions

Reviews

No rating value

Active Filters