Skip to Content
Merck
All Photos(1)

Key Documents

1225226

USP

Dopamine Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-Methoxytyramine hydrochloride, 3-MT, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

30 MG
€1,200.00

€1,200.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
30 MG
€1,200.00

About This Item

Linear Formula:
CH3OC6H3-4-(OH)CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
3631283
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€1,200.00


Estimated to ship on28 May 2025


Request a Bulk Order

API family

dopamine

packaging

pkg of 30 mg

manufacturer/tradename

USP

mp

213-215 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Cl[H].COc1cc(CCN)ccc1O

InChI

1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H

InChI key

AWRIOTVUTPLWLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Dopamine Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Dopamine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service