Skip to Content
Merck
All Photos(1)

Key Documents

Z688436

Lab-Sorb granules

absorbs aqueous or mixed aqueous and organic solvent waste

Synonym(s):

spill control, spill granules

Sign Into View Organizational & Contract Pricing

Select a Size

16 OZ
€42.70

€42.70


Check Cart for Availability


Select a Size

Change View
16 OZ
€42.70

About This Item

UNSPSC Code:
47131905
NACRES:
NB.35

€42.70


Check Cart for Availability

packaging

pack of 2 ea

manufacturer/tradename

Diversified Biotech SORB-1000

General description

Lab-Sorb reduces the possibility of accidentally spilling liquid waste in the lab. It is easy to use. The simple four step process takes less than two minutes. Lab-Sorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste.

Legal Information

Lab-Sorb is a trademark of Diversified Biotech

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service