Skip to Content
Merck
All Photos(1)

Key Documents

Z688444

Lab-Sorb starter kit

Variety Pack

Synonym(s):

spill control, spill granules

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€60.60

€60.60


Check Cart for Availability


Select a Size

Change View
1 KIT
€60.60

About This Item

UNSPSC Code:
47131905
NACRES:
NB.35

€60.60


Check Cart for Availability

feature

Contains 30 bags - 15 small, 10 medium, 5 large

manufacturer/tradename

Diversified Biotech SORB-2300

General description

Kit contains:
  • 16 oz tub of Lab-Sorb granules
  • 15 small bags
  • 10 medium bags
  • 5 large bags
Lab-Sorb reduces the possibility of accidentally spilling liquid waste in the lab. It is easy to use, the simple four step process takes less than two minutes. Lab-Sorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste.

Legal Information

Lab-Sorb is a trademark of Diversified Biotech

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I'm curious about if this is able to be autoclaved or if this could work with biohazardous waste for safe disposal of biohazardous liquids?

    1 answer
    1. One 16 oz. tub of Lab-Sorb will absorb up to 50 liters of liquid. The material may be autoclaved and will retain any liquid during the process.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service