Skip to Content
Merck
All Photos(1)

Key Documents

1.03874

Supelco

CHEMIZORB® H+

Sign Into View Organizational & Contract Pricing

Select a Size

2 KG
€111.00
5 KG
€246.00

€111.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
2 KG
€111.00
5 KG
€246.00

About This Item

UNSPSC Code:
47131902
NACRES:
NB.21

€111.00


Estimated to ship on24 May 2025


Request a Bulk Order

concentration

30-50% (acidimetric, Na2CO3)

loss

≤15% loss on drying, 160 °C

pH

9.0 - 12.0(5%, aqueous slurry)

bulk density

85 - 100 g/100 mL

suitability

passes test for application (Hydrochloric acid)
passes test for application (Nitric acid)
passes test for application (Sulfuric acid)
passes test for identity

General description

Absorbent and neutralizer for spilled acids, with indicator

Legal Information

CHEMIZORB is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service