Skip to Content
Merck
All Photos(2)

Key Documents

Z401099

Sigma-Aldrich

Biohazard spill kit

Disposable, L × W × D 7 1/4 in. × 6 1/2 in. × 5 1/4 in.

Synonym(s):

Biohazard Cleanup Kit, Hazardous Spill Kit, Spill Response Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€73.10

€73.10


Check Cart for Availability


Select a Size

Change View
1 EA
€73.10

About This Item

UNSPSC Code:
47131905
NACRES:
NB.35

€73.10


Check Cart for Availability

L × W × D

7 1/4 in. × 6 1/2 in. × 5 1/4 in.

Looking for similar products? Visit Product Comparison Guide

General description

Handles up to 2 gal spills of biohazardous materials. Disposable kit contains personal protective, spill pick up, and disposal items, and is packaged in a cardboard carton.

Contents:
1 x CPR clear mouth barrier
1 x plastic scoop
1 x pair nitrile gloves
1 x disposable, back fastening gown
1 x disposable dust mask with mouth shield
2 oz pack of congealing powder containing Potassium Polyacrylate
1 x disposable antiseptic towlette 3" x 6" active ingredient chloroxylrnol (0.5%)
1 x germicidal disposable wipe 5" x 8" active ingredients n-Alkyl dimethyl ethylbenzyl ammonium chloride (0.25%) & dimethyl benzyl ammonium chloride (0.25%)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service