Skip to Content
Merck
All Photos(1)

Key Documents

SRP5171

Sigma-Aldrich

Catenin α, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAP102, CTNNA1, FLJ36832

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€325.00

€325.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€325.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€325.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~123 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CTNNA1(1495)

General description

Catenin α is a novel actin-binding and -bundling protein. Catenin α is responsible for organizing and tethering actin filaments at the zones of E-cadherin-mediated cell-cell contact. Monomeric Catenin α can bind strongly to E-Cadherin-β-Catenin, whereas the dimer preferentially binds actin filaments. Different molecular conformations are associated with these different binding states, indicating that Catenin α is an allosteric protein. Catenin α directly regulates actin-filament organization by suppressing Arp2/3-mediated actin polymerization, likely by competing with the Arp2/3 complex for binding to actin filaments

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Rimm et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(19), 8813-8817 (1995-09-12)
Calcium-dependent homotypic cell-cell adhesion, mediated by molecules such as E-cadherin, guides the establishment of classical epithelial cell polarity and contributes to the control of migration, growth, and differentiation. These actions involve additional proteins, including alpha- and beta-catenin (or plakoglobin) and
Frauke Drees et al.
Cell, 123(5), 903-915 (2005-12-06)
Epithelial cell-cell junctions, organized by adhesion proteins and the underlying actin cytoskeleton, are considered to be stable structures maintaining the structural integrity of tissues. Contrary to the idea that alpha-catenin links the adhesion protein E-cadherin through beta-catenin to the actin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service