Skip to Content
Merck
All Photos(1)

Key Documents

SMB01004

Sigma-Aldrich

Chrysogine

≥90% (LC/MS-ELSD)

Synonym(s):

(+)-Chrysogine, 2-[(1R)-1-hydroxyethyl]-3H-quinazolin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€271.00

€271.00


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€271.00

About This Item

Empirical Formula (Hill Notation):
C10H10N2O2
CAS Number:
Molecular Weight:
190.20
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€271.00


Estimated to ship on11 April 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

190.2

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

n1c2c(c(nc1C(O)C)O)cccc2

InChI

1S/C10H10N2O2/c1-6(13)9-11-8-5-3-2-4-7(8)10(14)12-9/h2-6,13H,1H3,(H,11,12,14)

InChI key

BMBSGGZMJQTQSO-UHFFFAOYSA-N

General description

Chrysogine, a quinazolinone metabolite, is a yellow pigment commonly produced by Fusarium sambucinum, Penicillium chrysogenum, Penicillium notatum, and other filamentous fungi. Existing research indicates that this bioactive natural compound, exhibits various biological properties, including antibacterial, antifungal, and antiviral activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annarita Viggiano et al.
Applied and environmental microbiology, 84(4) (2017-12-03)
Chrysogine is a yellow pigment produced by Penicillium chrysogenum and other filamentous fungi. Although the pigment was first isolated in 1973, its biosynthetic pathway has so far not been resolved. Here, we show that deletion of the highly expressed nonribosomal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service