Skip to Content
Merck
All Photos(1)

Key Documents

SRP5172

Sigma-Aldrich

Catenin β, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CTNNB, FLJ25606

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€322.00

€322.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€322.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

€322.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~115 kDa

technique(s)

activity assay: suitable

solubility

water: soluble

suitability

suitable for molecular biology

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CTNNB1(1499)

General description

Research area: Apoptosis. β-catenins are cytoplasmic proteins that are ubiquitously expressed. These proteins are associated with E-cadherin at cellular junctions.[1] CTNNB1 (catenin β-1) belongs to the armadillo family of proteins. This protein possesses a 130 amino acid N-terminal domain, 12 imperfect repeats of 42 amino acids termed arm repeats, and a 100 amino acid C-terminal domain. [2] The gene encoding β-catenin is localized on human chromosome 3p22.1.[3]

Application

Catenin β, GST tagged human has been used:
  • as a component of protein kinase buffer for in vitro kinase assay[4]
  • in in vitro citrullination assay to characterize β-catenin citrullination[5]
  • to perform differentiation factor screening[6]

Biochem/physiol Actions

β-catenin interacts with T-cell factor and lymphoid enhancer-binding factor (TCF and LEF) transcription factors and is an essential member of the Wingless-Wnt signal transduction pathway.[1] The adenomatous polyposis coli (APC) tumor-suppressor protein, together with Axin and glycogen synthase kinase 3 β (GSK3β), form a Wnt-regulated signaling complex that mediates phosphorylation-dependent degradation of βcatenin by the proteasome.[2] APC and the Siah-1 E3 ubiquitin-protein ligase-1 mediate a novel β-catenin degradation pathway linking p53 activation to cell cycle control. Activating mutations in the human β-catenin gene have been found in human colon cancer and melanomas.[7][8] The protein has a role in homeostasis and development.[2]

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Qu et al.
Nature chemical biology, 14(1), 94-101 (2017-10-31)
Wnt (wingless)/β-catenin signaling is critical for tumor progression and is frequently activated in colorectal cancer as a result of the mutation of adenomatous polyposis coli (APC); however, therapeutic agents targeting this pathway for clinical use are lacking. Here we report
Spleen tyrosine kinase activity regulates epidermal growth factor receptor signaling pathway in ovarian cancer
Yu Y, et al.
EBioMedicine, 47, 184-194 (2019)
? catenin in health: A review
Sharada Prakash
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2015)
C Arbore et al.
Nature communications, 13(1), 1146-1146 (2022-03-05)
α-catenin is a crucial protein at cell junctions that provides connection between the actin cytoskeleton and the cell membrane. At adherens junctions (AJs), α-catenin forms heterodimers with β-catenin that are believed to resist force on F-actin. Outside AJs, α-catenin forms
Somatic mutations of the ?-catenin gene are frequent in mouse and human hepatocellular carcinomas
Alix de La Coste
Proceedings of the National Academy of Sciences of the USA (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service