Skip to Content
Merck
All Photos(1)

Key Documents

SAB2104747

Sigma-Aldrich

Anti-METTL3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-M6A, Anti-MGC4336, Anti-MT-A70, Anti-Spo8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€486.00

€486.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€486.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€486.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

human, mouse, rabbit, horse, rat, guinea pig, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... METTL3(56339)

General description

Methyltransferase like 3 (METTL3), an RNA methyltransferase, is a member of the class I methyltransferase (MTase) family and contains the MTase domain. The METTL3 gene is mapped to human chromosome 14q11.2.

Immunogen

Synthetic peptide directed towards the middle region of human METTL3

Application

Anti-METTL3 antibody produced in rabbit has been used in western blotting.[1]

Biochem/physiol Actions

Methyltransferase like 3 (METTL3) is part of methyltransferase systems that mediates the m6methyladenosine modifications. It exists as a heterodimeric complex with METTL14. An elevated expression of METTL3 is observed in clear cell renal cell carcinoma and rheumatoid arthritis (RA). Mutations in the METTL3 gene are implicated in autoimmune thyroid disease (AITD) and neuroblastoma. It may function as a tumor-suppresser in colorectal cancer.

Sequence

Synthetic peptide located within the following region: IVAEVRSTSHKPDEIYGMIERLSPGTRKIELFGRPHNVQPNWITLGNQLD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Wang et al.
Nature, 534(7608), 575-578 (2016-06-10)
Chemical modifications of RNA have essential roles in a vast range of cellular processes. N(6)-methyladenosine (m(6)A) is an abundant internal modification in messenger RNA and long non-coding RNA that can be dynamically added and removed by RNA methyltransferases (MTases) and
Jun Bian et al.
Journal of cellular and molecular medicine, 24(16), 9280-9286 (2020-07-03)
Neuroblastoma ranks as the most commonly seen and deadly solid tumour in infancy. The aberrant activity of m6 A-RNA methyltransferase METTL3 is involved in human cancers. Therefore, functional genetic variants in the METTL3 gene may contribute to neuroblastoma risk. In
Wenhui Zheng et al.
Frontiers in oncology, 9, 1403-1403 (2020-01-11)
Methyltransferase-like 3 (METTL3), a predominantly catalytic enzyme in the N6-methyladenosine (m6A) methyltransferase system, is dysregulated and plays a dual role (oncogene or tumor suppressor) in different human cancers. The expression and pro- or anticancer role of METTL3 in different cancers
Fang Yu et al.
Nucleic acids research, 49(10), 5779-5797 (2021-05-29)
Faithful genome integrity maintenance plays an essential role in cell survival. Here, we identify the RNA demethylase ALKBH5 as a key regulator that protects cells from DNA damage and apoptosis during reactive oxygen species (ROS)-induced stress. We find that ROS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service