Skip to Content
Merck
All Photos(1)

Documents

SCP0192

Sigma-Aldrich

MMP 2 Substrate, fluorogenic

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H68N14O15
Molecular Weight:
1093.15
UNSPSC Code:
12352200
NACRES:
NA.32

product name

MMP 2 Substrate, fluorogenic,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥73%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

MCA-Pro-Leu-Ala-Nva-DNP-Dap-Ala-Arg-NH2

General description

Matrix metalloproteinases (MMPs) are localized to the nucleus, mitochondria, vesicles and cytoplasm. MMP family comprises of 25 members, of which 24 are present in mammals. MMPs are encoded as inactive enzymes and are localized to cell membranes. All MMP members possess pro domain (80 amino acids) and catalytic domains.

Application

Matrix metalloproteinases (MMP) are a family of zinc-dependent endopeptidases that degrade matrix proteins. MMPs include collagenases, gelatinases, matrilysins, enamelysins, metalloelastases, stromelysins and other structural protein and receptor lysins. The activity of these enzymes is selectively inhibited or measured by appropriately labeled peptides. MCA-Pro-Leu-Ala-Nva-DNP-Dap-Ala-Arg-NH2; DNP-Pro-Leu-Gly-Met-Trp-Ser-Arg (DNP-PLGMWSR), Dnp-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg-NH2 (DNP-PLGLWA-D-R-NH2), Dnp-Pro-Gln-Gly-Ile-Ala-Gly-Gln-D-Arg-OH (DNP-PQGIAGQ-D-R-OH) and MOCAc [(7-methoxy coumarin-4-yl) acetyl]-RPKPYANvaWMK(Dnp[2,4-dinitrophenyl])-NH(2) are substrates for matrix metalloproteinase 2.

Biochem/physiol Actions

Matrix metalloproteinases (MMPs) are responsible for degrading membrane proteins, such as basal lamina proteins of cerebral blood vessels. Proteolysis of intracellular substrates by MMPs is associated with the innate immune system and apoptosis. They are also involved in disease conditions such as oncogenesis, cardiac, neurological and autoimmune disorders. MMPs are also associated with pathogenesis related to protein conformational changes. Elevated expression of a number of MMPs is observed in myocardial infarction and heart failure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix Metalloproteinase Biology, 11-11 (2015)
Matrix Metalloproteinases In Health And Disease: Sculpting The Human Body (2017)
Control of matrix metalloproteinase catalytic activity
Ra HJ and Parks WC
Matrix Biology, 26(8), 587-596 (2007)
Maria Orfanoudaki et al.
Marine drugs, 18(1) (2020-01-08)
Mycosporine-like amino acids (MAAs) are water-soluble metabolites, reported to exhibit strong UV-absorbing properties. They have been found in a wide range of marine organisms, especially those that are exposed to extreme levels of sunlight, to protect them against solar radiation.
Matrix metalloproteinase-mediated disruption of tight junction proteins in cerebral vessels is reversed by synthetic matrix metalloproteinase inhibitor in focal ischemia in rat
Yang YI
Journal of Cerebral Blood Flow and Metabolism, 27(4), 697-709 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service