Skip to Content
Merck
All Photos(1)

Key Documents

SCP0193

Sigma-Aldrich

MMP Substrate

≥95% (HPLC), lyophilized

Synonym(s):

FS-6

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€186.00

€186.00


Usually ships in 2 weeks.


Select a Size

Change View
1 MG
€186.00

About This Item

Empirical Formula (Hill Notation):
C55H80N16O16
Molecular Weight:
1221.32
UNSPSC Code:
12352200
NACRES:
NA.32

€186.00


Usually ships in 2 weeks.

Product Name

MMP Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

MCA-Lys-Pro-Leu-Gly-Leu-DNP-Dpa-Ala-Arg-NH2

General description

Matrix Metalloproteases (MMPs) belongs to the zinc metalloprotease family M10.[1] They are zinc-dependent, calcium-containing hydrolases.[2]

Biochem/physiol Actions

Matrix Metalloproteases (MMPs) are involved in extracellular matrix (ECM) degradation.[1] MMPs regulate cellular and disease processes. They are implicated in processes such as cell proliferation, migration, differentiation, angiogenesis, anti-inflammatory response, vasoconstriction, apoptosis and host defense. Dysregulation of MMP is associated with arthritis, ulcers, encephalomyelitis and cancer.[2]
The peptide MCA-Lys-Pro-Leu-Gly-Leu-DNP-Dpa-Ala-Arg-NH2, or FS-6, is a fluorogenic substrate for several matrix metalloproteinases (MMPs). FS-6 is a water-soluble, modified form of the well-established MMP substrate FS-1, where a lysine residue has been added to the N-terminus of FS-1. The resulting FS-6 has been reported to give a peptide with improved kinetic properties for several MMPs, including MMP-1, MMP-8, MMP-13, and MMP-14, compared to FS-1. This MMP substrate has also been described as a particularly useful substrate for the zinc metalloproteinase tumor necrosis factor-α converting enzyme (TACE or ADAM-17). FS-6 can be used to measure metalloproteinase activity in tissue culture conditions, for example, on the surface of viable cells in situ.[3][4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knowledge-transfer learning for prediction of matrix metalloprotease substrate-cleavage sites
Wang Y, et al.
Scientific Reports, 7(1), 5755-5755 (2017)
Ulf Neumann et al.
Analytical biochemistry, 328(2), 166-173 (2004-04-29)
Matrix metalloproteinases (MMPs) and the related tumor necrosis factor converting enzyme (TACE) are involved in tissue remodeling, cell migration, and processing of signaling molecules, such as cytokines and adhesion molecules. Fluorescence-quenched peptide substrates have been widely used to quantitate the
Hideaki Nagase et al.
Cardiovascular research, 69(3), 562-573 (2006-01-13)
Matrix metalloproteinases (MMPs), also called matrixins, function in the extracellular environment of cells and degrade both matrix and non-matrix proteins. They play central roles in morphogenesis, wound healing, tissue repair and remodelling in response to injury, e.g. after myocardial infarction
MMP-12 catalytic domain recognizes triple helical peptide models of collagen V with exosites and high activity.
Bhaskaran R
The Journal of Biological Chemistry, 283(31), 21779-21788 (2008)
David J Wilkinson et al.
The FEBS journal (2021-07-17)
Matrix metalloproteinase-13 (MMP-13) is a uniquely important collagenase that promotes the irreversible destruction of cartilage collagen in osteoarthritis (OA). Collagenase activation is a key control point for cartilage breakdown to occur, yet our understanding of the proteinases involved in this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service