Skip to Content
Merck
All Photos(1)

Key Documents

444221

Sigma-Aldrich

MMP-1/MMP-9 Substrate, Fluorogenic

An excellent quenched fluorescence substrate for MMP-1 and MMP-9.

Synonym(s):

MMP-1/MMP-9 Substrate, Fluorogenic

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€484.00

€484.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€484.00

About This Item

Empirical Formula (Hill Notation):
C49H68N14O12S
Molecular Weight:
1077.22
UNSPSC Code:
12352200
NACRES:
NA.77

€484.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

50% acetic acid: 10 mg/mL

fluorescence

λex ~365 nm
λem ~450 nm

shipped in

ambient

storage temp.

−20°C

General description

An excellent quenched fluorescence substrate for MMP-1 and MMP-9 (excitation maximum: 365 nm; emission maximum: 450 nm). Both enzymes cleave this substrate between Gly and Cys(Me), liberating a fluorescent cleavage product. Suitable for inhibitor screening and determining Ki values. Excitation max: ~365 nm; emission max: ~450 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMP-1, MMP-9
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

{DNP-Pro-Cha-Gly-Cys(Me)-His-Ala-Lys(N-Me-Abz)-NH₂[Cha = β-cyclohexylalanyl; Abz = 2-aminobenzoyl (anthraniloyl)]

Other Notes

Bickett, D.M., et al. 1993. Anal. Biochem. 212, 58.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service