Skip to Content
Merck
All Photos(1)

Key Documents

444212

Sigma-Aldrich

MMP-2 Substrate, Fluorogenic

A quenched fluorescence substrate for MMP-2 (gelatinase A; kcat/Km = 3.97 x 10⁵ M⁻¹s⁻¹).

Synonym(s):

MMP-2 Substrate, Fluorogenic

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€272.00

€272.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€272.00

About This Item

Empirical Formula (Hill Notation):
C49H68N14O15
Molecular Weight:
1093.15
UNSPSC Code:
12352200
NACRES:
NA.77

€272.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

fluorescence

λex ~325 nm
λem ~393 nm

shipped in

ambient

storage temp.

−20°C

General description

A quenched fluorescence substrate for MMP-2 (gelatinase A; kcat/Km = 3.97 x 105 M-1s-1; excitation maximum: ~325 nm; emission maximum: ~393 nm).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A quenched fluorescence substrate for MMP-2 (gelatinase A)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

MCA-Pro-Leu-Ala-Nva-Dpa-Ala-Arg-NH₂[Dpa = 3-(2,4-dinitrophenyl)-L-2,3-diaminopropionyl; Nva = L-norvaline]

Other Notes

Murphy, G., et al. 1994. J. Biol. Chem. 269, 6632.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service