Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Check Cart for Availability | € 360,00 |
About This Item
NACRES:
NA.85
UNSPSC Code:
41106610
€ 360,00
Check Cart for Availability
storage temp.
−20°C
Quality Segment
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



