Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
40 reactions | Check Cart for Availability | € 357,00 |
About This Item
NACRES:
NA.51
UNSPSC Code:
41106610
€ 357,00
Check Cart for Availability
form
solid
Quality Segment
manufacturer/tradename
Novagen®
storage condition
OK to freeze
shipped in
wet ice
storage temp.
−20°C
Features and Benefits
• No restriction enzymes or special primers
• Direct ligation of PCR product with vector
• Compatible with polymerases that leave single 3´-dA overhangs
• Blue/white screening with pSTBlue-1 or pETBlue-1 vectors
• Simple protocol takes as little as 40 minutes from PCR product to plating transformants
• Direct ligation of PCR product with vector
• Compatible with polymerases that leave single 3´-dA overhangs
• Blue/white screening with pSTBlue-1 or pETBlue-1 vectors
• Simple protocol takes as little as 40 minutes from PCR product to plating transformants
Other Notes
20 rxn40 rxn
•2 × 0.5 µg4 × 0.5 µgpETBlue-1 AccepTor Vector
•10 µl10 µlAccepTor Vector Positive Control Insert
•2 × 0.5 µg4 × 0.5 µgpETBlue-1 AccepTor Vector
•10 µl10 µlAccepTor Vector Positive Control Insert
Legal Information
This product is covered by U.S. Patent 5,856,144 owned by EMD Chemicals Inc. or its Affiliates.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form liquid | form liquid | form solid |
| storage temp. −20°C | storage temp. −20°C | storage temp. −70°C | storage temp. −20°C |
| shipped in wet ice | shipped in - | shipped in - | shipped in - |
| storage condition OK to freeze | storage condition OK to freeze | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition - |
| manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



