Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mg | Check Cart for Availability | € 549,00 € 466,65 |
About This Item
€ 466,65
List Price€ 549,00Save 15%biological source
goat
Quality Segment
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
mouse
technique(s)
neutralization: suitable, western blot: suitable
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Il10(16153)
General description
Anti-interleukin-10 antibody is specific for mouse IL-10. The antibody shows less than 5% cross reactivity to recombinant human IL-10.
Immunogen
Application
Biochem/physiol Actions
Physical form
Preparation Note
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form IgG fraction of antiserum | antibody form IgG fraction of antiserum | antibody form IgG fraction of antiserum | antibody form IgG fraction of antiserum |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source goat | biological source goat | biological source goat | biological source goat |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| Gene Information mouse ... Il10(16153) | Gene Information human ... IL11(3589) | Gene Information mouse ... Il1a(16175) | Gene Information human ... IL2(3558) |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| I5145-1MG | 04061826675403 |



