Skip to Content
Merck
All Photos(1)

Documents

I5270

Sigma-Aldrich

Anti-Interleukin-11 antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-IL-11

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL11(3589)

General description

Interleukin-11 (IL-11) is a pleotropic cytokine that belongs to the IL-6 family. The effects of IL-11 include hemopoiesis, macrophage differentiation and thrombopoiesis. Many cell types including osteoblasts, trophoblasts, fibroblasts, chondrocytes and a variety of tissue such as brain, gut, spinal cord and testis secrete IL-11. The cellular effects of IL-11 are mediated by a multimeric complex consisting of IL-11Rα and gp130Rβ subunits. The pathways activated by IL-11 include JAK/STAT, Ras/MAPK and Grb2 signaling. IL-11 plays a major role in inflammatory responses, immune responses by regulating the production of IgG by B cells and in tumorigenesis-related inflammation
Anti-Interleukin-11 antibody specifically reacts with human IL-11.

Specificity

The antibody neutralizes the biological activity of recombinant human IL-11.

Immunogen

recombinant human IL-11 expressed in Sf 21 insect cells.

Application

Anti-Human IL-11 is suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengyi Tang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 11(26), e2306348-e2306348 (2024-05-02)
Patients who have non-small cell lung cancer (NSCLC) with epidermal growth factor receptor (EGFR) mutations are more prone to brain metastasis (BM) and poor prognosis. Previous studies showed that the tumor microenvironment of BM in these patients is immunosuppressed, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service