Skip to Content
Merck

Skip To

D5928

6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline

Powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 335,00

About This Item

Empirical Formula (Hill Notation):
C15H15N3O
CAS Number:
Molecular Weight:
253.30
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
MDL number:

€ 335,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline,

assay

99.8% (HPLC)

Quality Segment

form

powder

color

orange

solubility

acetonitrile: 20 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

OCCN1CCCc2cc(CC(C#N)C#N)ccc12

InChI

1S/C15H17N3O/c16-10-13(11-17)8-12-3-4-15-14(9-12)2-1-5-18(15)6-7-19/h3-4,9,13,19H,1-2,5-8H2

InChI key

ANBPREOOKGEEDY-UHFFFAOYSA-N

Application

Fluorescent molecular rotor

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C6206C6331320609
color

orange

color

yellow

color

very dark blue

color

faint yellow

form

powder

form

powder

form

powder

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

solubility

acetonitrile: 20 mg/mL

solubility

DMSO: 17.5 mg/mL, clear, bright yellow

solubility

acetonitrile: 1 mg/mL, clear, blue

solubility

-

assay

99.8% (HPLC)

assay

≥98% (HPLC)

assay

≥85% (HPLC)

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A A Al-Majed
Journal of pharmaceutical and biomedical analysis, 21(4), 827-833 (2000-03-04)
A simple colorimetric method for the determination of D-penicillamine in pure form and pharmaceutical formulations is described. The method is based on coupling between D-penicillamine and 2,6-dichloroquinone-4-chlorimide (DCQ) in dimethylsulphoxide. The optimum conditions for the reaction were investigated and incorporated
T Iio et al.
Journal of biochemistry, 113(2), 196-199 (1993-02-01)
1-(2-Hydroxyethyl)-6-[(2,2-dicyano)vinyl]-2,3,4-trihydroquinoli ne (DCQ) is a fluorescent dye and its fluorescence yield is determined by a freedom of intramolecular rotation. When DCQ was added to an actin solution, its fluorescence intensity increased as the actin molecule transformed from G-actin to F-actin.
T Iwaki et al.
Biochemistry, 32(29), 7589-7592 (1993-07-27)
We have prepared monoclonal antibodies for the fluorescent molecular rotors 9-(2-carboxy-2-cyanovinyl)julolidine (CCVJ) and 9-(dicyanovinyl)julolidine (DCVJ). Mouse monoclonal antibody (IgG2b) prepared against CCVJ-conjugated bovine serum albumin strongly bound CCVJ and DCVJ. The CCVJ (or DCVJ) binding to IgG and Fab was



Global Trade Item Number

SKUGTIN
D5928-5MG04061831998757

Questions

Reviews

No rating value

Active Filters