Skip to Content
Merck
All Photos(2)

Key Documents

320609

Sigma-Aldrich

2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate

powder

Synonym(s):

2,4-DCPD, 2,4-Dichlorophenyldiazonium 1,5-naphthalenedisulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€517.00

€517.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 G
€517.00

About This Item

Linear Formula:
Cl2C6H3N2(O3SC10H6SO3H) · xH2O
CAS Number:
Molecular Weight:
461.30 (anhydrous basis)
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€517.00


Estimated to ship on02 June 2025


Request a Bulk Order

form

powder

Quality Level

composition

Carbon: 37.5-45.8% (Anhydrous Basis)

Nitrogen: 5.0-7.0% (Anhydrous Basis)

Water: 3.0-10.0% (by Karl Fischer)

color

faint yellow

mp

195 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

O.Clc1ccc([N+]#N)c(Cl)c1.OS(=O)(=O)c2cccc3c(cccc23)S([O-])(=O)=O

InChI

1S/C10H8O6S2.C6H3Cl2N2.H2O/c11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16;7-4-1-2-6(10-9)5(8)3-4;/h1-6H,(H,11,12,13)(H,14,15,16);1-3H;1H2/q;+1;/p-1

InChI key

SPGAUZWLXGQOLT-UHFFFAOYSA-M

General description

2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate is a diazonium salt intended for use in histology and hematology.

Application

2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate is used in spectrophotometric detection of total bilirubin in serum.
Used in spectrophotometric detection of total bilirubin in serum.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service