Skip to Content
Merck

Skip To

D2154

5,6-Dehydroarachidonic acid

≥98%, ethanol solution

Synonym(s):

5,6-dehydro AA, cis-8,11,14-Eicosatrien-5-ynoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 369,00

About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352211
EC Number:
200-578-6
MDL number:

€ 369,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98%

form

ethanol solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CC#CCCCC(O)=O

InChI

1S/C20H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-

InChI key

GIOQWSLKUVKKAO-QNEBEIHSSA-N

General description

An analog of arachidonic acid in which the 5,6 double bond has been replaced with an acetylene group.

Biochem/physiol Actions

Inhibits 5-lipoxygenase of guinea pig leukocytes with an IC50 of 10 μM. In extracts of rat basophilic leukemia cells preincubated with 5,6-dehydro AA, the Ki for the conversion of arachidonic acid to 5-HPETE is 15 μM.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
900411E181198735000
assay

≥98%

assay

>99% (TLC)

assay

≥99% (TLC and GC)

assay

≥97% (CP)

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C

form

ethanol solution

form

ethanol solution

form

oil

form

liquid

Quality Level

200

Quality Level

-

Quality Level

100

Quality Level

200


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

62.6 °F - closed cup

flash_point_c

17 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



H H Tai et al.
Biochemistry, 19(9), 1989-1993 (1980-04-29)
[5,6-3H]arachidonic acid has been prepared by catalytic reduction of eicosa-cis-8,11,14-trien-5-ynoic acid (IV) over Lindlar catalyst. When either [5,6-3H]arachidonic acid or [5,6-3H]PGH2 is converted into PGI2 by swine aortic microsomes, the tritium at C-6 is lost to the medium. Thus, the
Strong suppression by mononuclear leukocytes from human newborns on maternal leukocytes: mediation by prostaglandins.
S A Johnsen et al.
Clinical immunology and immunopathology, 23(3), 606-615 (1982-06-01)
E Razin et al.
Journal of immunology (Baltimore, Md. : 1950), 133(2), 938-945 (1984-08-01)
The quantitative relationships between the secretion of a granule-associated mediator, beta-hexosaminidase, and the oxidative metabolism of arachidonic acid by the 5-lipoxygenase pathway were analyzed for a homogeneous population of T cell-dependent, bone marrow-derived, murine mast cells. The mast cells were



Global Trade Item Number

SKUGTIN
D2154-25UG04061838070678
D2154-100UG04061837896828