Skip to Content
Merck
All Photos(1)

Key Documents

900411E

Avanti

arachidonic acid-alkyne

Avanti Research - A Croda Brand 900411E, ethanol solution

Synonym(s):

5Z,8Z,11Z,14Z-eicosatetraen-19-ynoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€506.00

€506.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€506.00

About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€506.00


Check Cart for Availability

Request a Bulk Order

form

ethanol solution

packaging

pkg of 1 × 1 mL (900411E-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900411E

concentration

1 mg/mL (900411E-1mg)

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

General description

Arachidonic acid (ARA) is a polyunsaturated ω-6 fatty acid. It is composed of 20-carbon chain fatty acid. ARA is found in fish, eggs, poultry and sea food. It is present abundantly in phospholipids of cell membranes.[1]

Biochem/physiol Actions

Arachidonic acid (ARA) governs the cell membrane fluidity and ion channels activity. Accumulation of free ARA leads to cytotoxicity followed by cell death.[1]

Packaging

5 mL Amber Screw Cap Bottle with Foil Bag Argon Overseal (900411E-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arachidonic acid: physiological roles and potential health benefits-a review
Tallima H and El Ridi R
Journal of Advanced Research, 11, 33-41 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service