CLS432006
Corning® CoolCell™ FTS30
freezing container, (30 x 1 mL or 2 mL cryogenic vials), purple
Synonym(s):
cryovials
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
foamed PE
feature
Holds 30 x 1-2 mL cryovials
packaging
pkg of 1
manufacturer/tradename
Corning 432006
technique(s)
cryopreservation: suitable
H × diam.
11.5 cm × 16.5 cm , 12.7mm well diameter
color
purple
capacity
(30 x 1 mL or 2 mL cryogenic vials)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
CoolCelll FTS30 utilizes a solid state core and controlled microconvection technology to evenly draw in -80°C freezer air through a bottom base vent, uniformly disperse the cold air around each vial in the central chamber and then release the thermal load from the vials through a top vent choke. The inner vial module holds 30 cryogenic vials and can be removed in one step. Each vial achieves a uniform and reproducible -1°C/minute freezing profile and thermal profiles are highly reproducible. Due to the low thermal mass of the uniform-density cross-linked polyethylene foam container, freezing can be conducted without a rise in local freezer temperature, thereby protecting nearby samples.
- For 30 standard 1.0 mL to 2.0 mL cryogenic vials.
- Controlled micro-convection for uniform freezing of 30 vials.
- Removable vial tray for one-step transfer of samples into and out of freezing chamber.
Legal Information
CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service