Skip to Content
Merck
All Photos(1)

Key Documents

CLS432000

Corning® CoolCell Freezer Container

purple

Synonym(s):

cryovials

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€167.00

€167.00


Check Cart for Availability


Select a Size

Change View
1 EA
€167.00

About This Item

UNSPSC Code:
41123300
NACRES:
NC.01

€167.00


Check Cart for Availability

material

foamed PE

feature

Holds 12 x 1-2 mL cryovials

packaging

pkg of 1

manufacturer/tradename

Corning 432000

technique(s)

cryopreservation: suitable

color

purple

General description

Corning® CoolCell freezer containers are ideal for cryopreservation of stem cells, primary cells, PBMC, cell lines, insect cells, yeast, and others. The proprietary Corning CoolCell technology utilizes a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation.

Features and Benefits

  • Corning® CoolCell container, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute
  • Easy to use with quick re-use time
  • Inexpensive than alcohol-based devices and no ongoing cost involved
  • Available in purple, green, orange, and pink
  • Radially symmetric for all vials to have a uniform freeze rate
  • Numbered wells for easy sample identification
  • Designed with a beveled lid for steady grip and easy opening
  • Vial tips can be removed by tipping the device upside down

Legal Information

CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service