Skip to Content
Merck
All Photos(1)

Key Documents

CLS3615

Corning® 96 Well Special Optics Microplate

flat bottom clear, polystyrene, bag of 25, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates, optically clear flat bottom plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€1,580.00

€1,580.00


Check Cart for Availability


Select a Size

Change View
100 EA
€1,580.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

€1,580.00


Check Cart for Availability

material

black/clear bottom polystyrene plate
clear bottom
flat bottom clear
polystyrene

sterility

non-sterile

feature

lid: no
lid: no
skirt
plate format: 96 well special optics

packaging

case of 100
bag of 25

manufacturer/tradename

Corning 3615

maximum volume

360 μL

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

working volume

200 μL

color

black

suitability

suitable for (optically clear flat well bottom permits direct microscopic viewing)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 96 Well Special Optics Plates

The 96 well special optics plates have black, opaque walls, and ultra thin flat clear bottoms that permit direct microscopic viewing and improved optical performance in fluorescent-based assays.
  • Special optics 96 well microplates have black walls with ultra thin, clear bottoms for sharp, clear images and minimal background in fluorescent assays
  • Designed to reduce well-to-well crosstalk during fluorescent assays
  • Can be used for both top and bottom reading instrument
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
  • Total volume is 360μL; recommended working volume of 75-200μL
  • Conforms to standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service