Skip to Content
Merck
All Photos(1)

Key Documents

CLS3904

Corning®  96 Well Black Polystyrene Microplate

flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

96 mulitwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€1,180.00

€1,180.00


Check Cart for Availability


Select a Size

Change View
100 EA
€1,180.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black polystyrene
clear bottom
flat bottom clear
polystyrene
size:
96 wells

€1,180.00


Check Cart for Availability

material

black polystyrene
clear bottom
flat bottom clear
polystyrene

sterility

sterile

feature

lid
skirt
plate format: 96 well standard

packaging

case of 100
bag of 20

manufacturer/tradename

Corning 3904

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

well volume

360 μL

well working volume

75-200 μL

color

black

suitability

suitable for (fluorescent/luminescent assays)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Black Polystyrene Microplate have opaque black polystyrene walls to minimize well-to-well crosstalk and background fluorescence and/or luminescence.
  • Optically clear flat well bottom permits direct microscopic viewing.
  • Applicable for both top and bottom reading instruments.
  • Nonreversible lids with condensation ring to reduce contamination.
  • Non-Pyrogenic and sterilized by gamma radiation.
  • Hydrophilic and negatively charged surface optimized for cell attachment and growth.
  • Product does not contain materials of animal origin.
  • Tested for the attribute of cell attachment and growth utilizing an attachment-dependent mammalian cell line.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer H R Kenty et al.
PloS one, 10(4), e0122044-e0122044 (2015-04-09)
Studying the response of islet cells to glucose stimulation is important for understanding cell function in healthy and disease states. Most functional assays are performed on whole islets or cell populations, resulting in averaged observations and loss of information at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service