Skip to Content
Merck
All Photos(1)

Key Documents

CLS3614

Corning® 96 Well Special Optics Microplate

flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 25, sterile

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates, optically clear flat bottom plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€1,670.00

€1,670.00


Check Cart for Availability


Select a Size

Change View
100 EA
€1,670.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

€1,670.00


Check Cart for Availability

material

black polystyrene
black/clear bottom polystyrene plate
clear bottom
flat bottom clear

sterility

sterile

feature

lid: no
skirt
plate format: 96 well special optics

packaging

case of 100
bag of 25

manufacturer/tradename

Corning 3614

maximum volume

360 μL

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

working volume

200 μL

suitability

suitable for (optically clear flat well bottom permits direct microscopic viewing)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 96 Well Special Optics Plates

The 96 well special optics plates have black, opaque walls, and ultra thin flat clear bottoms that permit direct microscopic viewing and improved optical performance in fluorescent-based assays.
  • Ultra thin, optically clear flat well bottom permits direct microscopic viewing, resulting in lower background fluorescence and enabling readings down to 340nm
  • Opaque walls to prevent well-to-well crosstalk
  • Can be used for both top and bottom reading instruments
  • Flat bottoms with 360μL total volume
  • Recommended working volumes of 75 to 200μL
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Without lids
  • Bulk packed 25 per bag

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan D Trinklein et al.
mAbs, 11(4), 639-652 (2019-01-31)
T-cell-recruiting bispecific antibodies (T-BsAbs) have shown potent tumor killing activity in humans, but cytokine release-related toxicities have affected their clinical utility. The use of novel anti-CD3 binding domains with more favorable properties could aid in the creation of T-BsAbs with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service