Skip to Content
Merck
All Photos(1)

Key Documents

CLS3098

Corning® multiwell plates, plate lids and sealing mats

universal multiwell plate lid w/o corner notch, polystyrene, clear, sterile, 100/cs

Synonym(s):

universal microplate lid, universal microtiter plate, universal plate lid

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€313.00

€313.00


Check Cart for Availability


Select a Size

Change View
100 EA
€313.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
polystyrene
size:
96 or 384 wells

€313.00


Check Cart for Availability

material

polystyrene

sterility

sterile

feature

lid
skirt (half skirt)

packaging

case of 100
pack of 25

manufacturer/tradename

Corning 3098

size

96 or 384 wells

color

clear

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

General description

  • Lids are made of rigid polystyrene
  • Lids do not have a corner notch on the A1 corner and does not need to be oriented in any particular direction to be placed on Corning® plates
  • Lid has a shorter skirt than standard lids
  • Lids do not fit Corning 2 μL 1536 well microplates
  • Suitable for 96 well, 96 half area & 384 well plates
  • Sterilized by gamma radiation

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service