Skip to Content
Merck
All Photos(1)

Documents

CLS3922

Corning® 96 well plates

96 well plate, polystyrene, High Bind, white flat bottom wells, non-sterile, white, 100/cs

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

flat bottom wells white
polystyrene
white polystyrene plate

sterility

non-sterile

feature

lid: no
skirt
plate format: 96 well standard

packaging

case of 100
pack of 25

manufacturer/tradename

Corning 3922

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

white

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • White microplates enhance luminescent signals and have low background luminescence and fluorescence
  • Designed to reduce well-to-well crosstalk
  • High binding surface is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Binding capacity is approximately 400 to 500ng IgG/cm2
  • Ideal for immunoassays
  • Without lids, lids supplied separately
  • Total volume is 360μL; recommended working volume of 75 to 200μL
  • Conforms to standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service