Key Documents
BR782151
BRAND® lids for microplates BRAND®plates
plate size 96 wells, without condensation rings
Synonym(s):
Accessories, BRANDplates®, Lids, Microplates
Select a Size
100 EA
€85.50
€85.50
Check Cart for Availability
Select a Size
Change View
100 EA
€85.50
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
polystyrene
€85.50
Check Cart for Availability
material
polystyrene
packaging
pack of 100 ea
manufacturer/tradename
BRAND 782151
plate size
96 wells
suitability
suitable for (bacteriological assays)
General description
To protect samples, reaction batches or cell cultures from both contamination and evaporation, compatible lids are available for all BRANDplates® microplates. The lids are made of crystal-clear polystyrene. They are optimally matched to the corresponding BRANDplates® microplates, so that plates with lids can also be stacked. The cut-corner sets the orientation of the lid. For 96-well standard plates, lids with condensation rings are available.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)