Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 10 ea | Check Cart for Availability | € 184,00 |
About This Item
NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polypropylene screw closure, retained silicone gasket
Packaging:
pack of 10 ea
€ 184,00
Check Cart for Availability
material
polypropylene screw closure, retained silicone gasket
packaging
pack of 10 ea
manufacturer/tradename
Nalgene DS3132-0024
diam.
24 mm
General description
Ensure leakproof service of Nalgene™ of Oak Ridge centrifuge bottles and tubes at their recommended maximum rated speeds by using these specific sealing caps, also recommended for use when spinning hazardous materials.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
1 of 1
This Item | |||
|---|---|---|---|
| material polypropylene screw closure, retained silicone gasket | material polypropylene screw closure, retained silicone gasket | material polypropylene screw closure, retained silicone gasket | material polypropylene cap, red cap |
| manufacturer/tradename Nalgene DS3132-0024 | manufacturer/tradename Nalgene DS3132-0020 | manufacturer/tradename Nalgene DS3132-0063 | manufacturer/tradename Nalgene 362150-5240 |
| packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 2 ea | packaging case of 1000 ea |
| diam. 24 mm | diam. 20 mm | diam. 63 mm | diam. - |
Still not finding the right product?
Explore all of our products under Nalgene® sealing caps
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C7437-10EA | 04061832614533 |

