Skip to Content
Merck

Skip To

C7312

Nalgene® sealing caps

diam. 20 mm, for use with 28 and 30 mL tubes, pack of 10 ea

Synonym(s):

bottle cap, bottle closure, Nalgene DS3132-0020, bottle seal, plastic cap

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 ea
Check Cart for Availability
€ 203,00

About This Item

NACRES:
NB.17
UNSPSC Code:
41121703
Material:
polypropylene screw closure, retained silicone gasket
Packaging:
pack of 10 ea

€ 203,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene screw closure, retained silicone gasket

packaging

pack of 10 ea

manufacturer/tradename

Nalgene DS3132-0020

diam.

20 mm

General description

Ensure leakproof service of Nalgene™ of Oak Ridge centrifuge bottles and tubes at their recommended maximum rated speeds by using these specific sealing caps, also recommended for use when spinning hazardous materials.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7687C7437TMO362150-5240
material

polypropylene screw closure, retained silicone gasket

material

polypropylene screw closure, retained silicone gasket

material

polypropylene screw closure, retained silicone gasket

material

polypropylene cap, red cap

manufacturer/tradename

Nalgene DS3132-0020

manufacturer/tradename

Nalgene DS3132-0063

manufacturer/tradename

Nalgene DS3132-0024

manufacturer/tradename

Nalgene 362150-5240

packaging

pack of 10 ea

packaging

pack of 2 ea

packaging

pack of 10 ea

packaging

case of 1000 ea

diam.

20 mm

diam.

63 mm

diam.

24 mm

diam.

-


Still not finding the right product?

Explore all of our products under Nalgene® sealing caps




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
C7312-10EA04061832614526