Skip to Content
Merck

Skip To

C-28052

Dendritic Cell Generation Medium DXF

Ready-to-use kit including Basal Medium and SupplementMix, 250 ml

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mL
Check Cart for Availability
€ 915,00

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207

€ 915,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

packaging

pkg of 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

DC Generation Medium DXF (Ready-to-use)

Application

The Dendritic Cell (DC) Generation Medium DXF has been developed for the easy and efficient generation of immature as well as fully mature myeloid Dendritic Cells from peripheral blood monocytes. The DC Generation Medium DXF is a chemically defined and xeno-free formulation for use with freshly isolated cells. Due to the utilization of exclusively synthetic, recombinant or plant-sourced materials, this medium is free of all animal-derived components and substances of human origin with Human Serum Albumin as the only exception. The medium consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium. For freshly isolated cells, the Monocyte Attachment Medium (C-28051) is also needed.Recommended for:Human Monocytes (hMo), freshly isolated and cryopreservedHuman Mononuclear Cells (hMNC), freshly isolated

Preparation Note

Click here for more information.
Store the Basal Medium at 4 to 8°C in the dark, store the SupplementMix at -20°C immediately after arrival. Keep the Cytokine Pack at -20°C for long-term storage or at 4 to 8°C for up to 2 weeks. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Complete Medium supplemented with cytokines should be used within 2 weeks. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Analysis Note

All lots of PromoCell DC Media are subjected to comprehensive quality control tests using human peripheral blood mononuclear cells. Each lot of PromoCell DC Media is tested for the ability to support DC maturation verified by morphological evaluation and flow cytometry analysis. Each lot of PromoCell Monocyte Attachment Medium is tested for its ability to support optimal attachment and viability of peripheral blood Monocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-28054C-28019C-28050
form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

packaging

pkg of 250 ml

packaging

pkg of 250 ml

packaging

pkg of 500 ml

packaging

pkg of 250 ml

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters