Skip to Content
Merck
All Photos(1)

Key Documents

C-28054

Sigma-Aldrich

Dendritic Cell Generation Medium DXF

Basal Medium, 250 ml

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€247.00

€247.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€247.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

€247.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

DC Base Medium DXF (supplements sold separately)

Application

The Dendritic Cell (DC) Base Medium DXF is a user-customizable version of the DC Generation Media DXF that does not include cytokines. It must be adequately supplemented by the user for the generation of immature as well as fully mature myeloid Dendritic Cells from peripheral blood monocytes.Note: DC Base Medium DXF must be supplemented with additional cytokines. For freshly isolated cells, the Monocyte Attachment Medium (C-28051) is also needed.Recommended for:Human Monocytes (hMo), freshly isolated and cryopreservedHuman Mononuclear Cells (hMNC), freshly isolated

Quality

All lots of PromoCell DC Media are subjected to comprehensive quality control tests using human peripheral blood mononuclear cells. Each lot of PromoCell DC Media is tested for the ability to support DC maturation verified by morphological evaluation and flow cytometry analysis. Each lot of PromoCell Monocyte Attachment Medium is tested for its ability to support optimal attachment and viability of peripheral blood Monocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Complete Medium supplemented with cytokines should be used within 2 weeks. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.

Protocols

Generate mature human dendritic immune cells from PBMCs or CD34+ stem cell derived monocytes in 7 days in defined and serum-free dendritic cell culture media. Explore over 350 PromoCell products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service