Skip to Content
Merck

Skip To

A8356

ARP 101

≥98% (HPLC), solid

Synonym(s):

(2R)-2-[([1,1′-Biphenyl]-4-ylsulfonyl)(1-methylethoxy)amino]-N-hydroxy-3-methyl-butanamide, (R)-N-Hydroxy-2-(N-isopropoxybiphenyl-4-ylsulfonamido)-3-methylbutanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 236,00

About This Item

Empirical Formula (Hill Notation):
C20H26N2O5S
CAS Number:
Molecular Weight:
406.50
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid

€ 236,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

solubility

DMSO: >20 mg/mL, H2O: insoluble

storage temp.

−20°C

SMILES string

CC(C)ON([C@H](C(C)C)C(=O)NO)S(=O)(=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C20H26N2O5S/c1-14(2)19(20(23)21-24)22(27-15(3)4)28(25,26)18-12-10-17(11-13-18)16-8-6-5-7-9-16/h5-15,19,24H,1-4H3,(H,21,23)/t19-/m1/s1

InChI key

DGZZVIWCMGVHGV-LJQANCHMSA-N

Application

ARP 101 is a MMP-2 selective inhibitor. ARP 101 has diverse physiological roles including wound healing, cancer metastasis, angiogenesis, tissue remodeling in development, and embryogenesis.

Biochem/physiol Actions

ARP 101 is a MMP-2 selective inhibitor (Gelatinase A, EC 3.4.24.24; TBE-1; LG4A; MONA; Type IV Collagenase). ARP 101 has diverse physiological roles including wound healing, cancer metastasis, angiogenesis, tissue remodeling in development and embryogenesis. IC50 = 0.81 nM on MMP-2.
ARP 101 is a MMP-2 selective inhibitor.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2883S5192B0186
form

solid

form

powder

form

solid

form

solid

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥97% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: >20 mg/mL, H2O: insoluble

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: soluble ~28 mg/mL, H2O: insoluble

solubility

DMSO: >20 mg/mL


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters