Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | € 96,40 |
5 mg | Check Cart for Availability | € 318,00 |
About This Item
Empirical Formula (Hill Notation):
C23H25N3O4S2
CAS Number:
Molecular Weight:
471.59
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥97% (HPLC)
Form:
solid
assay
≥97% (HPLC)
form
solid
color
off-white
solubility
DMSO: >20 mg/mL
storage temp.
−20°C
SMILES string
COc1ccc(cc1)S(=O)(=O)NCCCC(=O)Nc2ccc(Sc3ccc(N)cc3)cc2
InChI
1S/C23H25N3O4S2/c1-30-19-8-14-22(15-9-19)32(28,29)25-16-2-3-23(27)26-18-6-12-21(13-7-18)31-20-10-4-17(24)5-11-20/h4-15,25H,2-3,16,24H2,1H3,(H,26,27)
InChI key
LCFUJBSKPDPGKO-UHFFFAOYSA-N
Biochem/physiol Actions
BI-6C9 is a tBid inhibitor and antiapoptotic.
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form powder | form solid | form solid |
| assay ≥97% (HPLC) | assay ≥98% (HPLC) | assay >98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C |
| solubility DMSO: >20 mg/mL | solubility DMSO: 20 mg/mL, clear | solubility DMSO: soluble 10 mg/mL | solubility DMSO: soluble >5 mg/mL at 60 °C |
| color off-white | color white to beige | color off-white | color white |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



