Skip to Content
Merck

Skip To

09014

DAR-1

BioReagent, suitable for fluorescence, ≥97.0% (HPCE)

Synonym(s):

4,5-Diamino-N,N,N′,N′-tetraethylrhodamine, 4,5-Diamino-rhodamine B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 316,00

About This Item

Empirical Formula (Hill Notation):
C28H32N4O3
CAS Number:
Molecular Weight:
472.58
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.32
MDL number:

€ 316,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥97.0% (HPCE)

form

powder

fluorescence

λex 566 nm; λem 586 nm in 0.1 M phosphate pH 7.4, NO

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2c(Oc3cc(ccc3C24OC(=O)c5c(N)c(N)ccc45)N(CC)CC)c1

InChI

1S/C28H32N4O3/c1-5-31(6-2)17-9-11-19-23(15-17)34-24-16-18(32(7-3)8-4)10-12-20(24)28(19)21-13-14-22(29)26(30)25(21)27(33)35-28/h9-16H,5-8,29-30H2,1-4H3

InChI key

LFXDWSUQAVSSJH-UHFFFAOYSA-N

Application

suitable as fluorescent NO-sensor

Other Notes

Sensitive fluorescent indicator for nitric oxide, NO, based on rhodamine chromophore; LOD in the region of 10 nM; it is more photostable and emits at longer wave-length than the corresponding derivative of fluorescein (DAF, Cat. No. 41524)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
906344927082319856
form

powder

form

granular

form

powder

form

-

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

200

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

assay

≥97.0% (HPCE)

assay

-

assay

≥95%

assay

99% (CP)

suitability

suitable for fluorescence

suitability

-

suitability

-

suitability

-

product line

BioReagent

product line

-

product line

-

product line

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the solution stability of chemical 09014 DAR-1?

    1 answer
    1. The product is soluble in DMSO at a concentration of 5mM and is stable for at least 2 years after receipt when stored at -20°C.

      Helpful?

Reviews

No rating value

Active Filters