Skip to Content
Merck
All Photos(2)

Key Documents

927082

Sigma-Aldrich

Ni(COD)(Cy-BQ)

greener alternative

≥95%

Synonym(s):

(1,5-cyclooctadiene)([1,1′:4′,1″-tercyclohexane]-3′,6′-diene-2′,5′-dione)nickel(0)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€176.00
500 MG
€552.00

€176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€176.00
500 MG
€552.00

About This Item

Empirical Formula (Hill Notation):
C26H36NiO2
Molecular Weight:
439.26
UNSPSC Code:
12352200
NACRES:
NA.21

€176.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Ni(COD)(Cy-BQ) is a bench-stable 18-electron nickel–olefin complex that can be used for a variety of reactions.

Reference

Structurally Diverse Bench-Stable Nickel(0) Pre-Catalysts: A Practical Toolkit for In Situ Ligation Protocols

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service