Z804789
UV Spec Qualification Kit
certified reference material
About This Item
Recommended Products
grade
certified reference material
manufacturer/tradename
Jaytee Biosciences Ltd (CRM-2100)
technique(s)
UV/Vis spectroscopy: suitable
application(s)
food and beverages
petroleum
pharmaceutical
plastics
format
neat
General description
Contains qualification standards for
- UV Photometric Accuracy
- Resolution
- Stray Light
This UV Spec Qualification Kit has been developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.
All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.
Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034
Application
Features and Benefits
The kit includes the below standards and blanks:
- Stray light
- Stray light blank
- Resolution
- Resolution blank
- Photometric accuracy blank
- Photometric accuracy 600mg/L
- Photometric accuracy 60mg/L
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system, Nervous system
Storage Class Code
3 - Flammable liquids
Flash Point(F)
71.6 °F
Flash Point(C)
22 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service