Skip to Content
Merck
All Photos(1)

Key Documents

487090

Sigma-Aldrich

Water-18O

99 atom % 18O

Synonym(s):

18O Labeled water

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€1,760.00

€1,760.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€1,760.00

About This Item

Linear Formula:
H218O
CAS Number:
Molecular Weight:
20.02
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

€1,760.00


Check Cart for Availability

Request more information

description

Normalized with respect to hydrogen

Quality Level

isotopic purity

99 atom % 18O

form

liquid

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.11 g/mL at 20 °C (lit.)

mass shift

M+2

storage temp.

room temp

SMILES string

[18OH2]

InChI

1S/H2O/h1H2/i1+2

InChI key

XLYOFNOQVPJJNP-NJFSPNSNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

H2180 is isotopically modified compound consisting of two hydrogen atoms covalently bonded to oxygen-18. It is most convenient to store, transport and use Oxygen-18 in the form of water enriched with this isotope.

Application

O-18 enriched water is used in Positron-Emission Tomography (PET), a nuclear imaging technique that is used to observe metabolic processes in the body as an aid to the diagnosis of disease such as tumors, staging of cancer, metastasis, dementia, etc. H2180 is currently the only potential universal bacterial substrate suitable for stable isotope probing (SIP). It has been used in the quantification of population-specific growth dynamics in benthic bacterial communities in response to changing redox conditions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H H Coenen et al.
Nuclear medicine and biology, 37(7), 727-740 (2010-09-28)
Positron emission tomography (PET) is a rapidly expanding clinical modality worldwide thanks to the availability of compact medical cyclotrons and automated chemistry for the production of radiopharmaceuticals. There is an armamentarium of fluorine-18 ((18)F) tracers that can be used for
Elizabeth A Rettedal et al.
MicrobiologyOpen, 4(2), 208-219 (2015-02-05)
Current limitations in culture-based methods have lead to a reliance on culture-independent approaches, based principally on the comparative analysis of primary semantides such as ribosomal gene sequences. DNA can be remarkably stable in some environments, so its presence does not
Ömer K Coskun et al.
The ISME journal, 13(6), 1546-1559 (2019-02-21)
The benthos in estuarine environments often experiences periods of regularly occurring hypoxic and anoxic conditions, dramatically impacting biogeochemical cycles. How oxygen depletion affects the growth of specific uncultivated microbial populations within these diverse benthic communities, however, remains poorly understood. Here

Questions

  1. Mit welcher analytischen Methode wird die Isotopenreinheit geprüft?

    1 answer
    1. The method to determine isotopic purity is Mass Spectrometry. This product is an M-Clarity level 200 item. The analytical methods are considered proprietary. Please see the link below to a sample Certificate of Analysis that notes they method:
      https://www.sigmaaldrich.com/certificates/COFA/48/487090/487090-BULK_______MBBC8728__.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service