Skip to Content
Merck
All Photos(1)

Key Documents

Z804908

Wavelength, Stray Light and Resolution Kit

certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€359.00

€359.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€359.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.21

€359.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

manufacturer/tradename

Jaytee Biosciences Ltd (CRM-2102)

application(s)

general analytical

General description

Contains qualification standards for
  • UV Resolution
  • UV Stray Light
  • UV Multi-Wavelength

Application

Wavelength, Stray Light, and Resolution Kit contain UV multi-wavelength standards for use in the qualification of HPLC UV detectors.

Features and Benefits

  • Manufactured and verified in an ISO 17025 accredited laboratory as per UKAS (United Kingdom Accreditation Service)
  • Supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification
  • Traceable instrument qualification adds confidence to all data generated
  • Accompanied with a certificate of analysis (COA) having information on the expiry date for an effective audit

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Met. Corr. 1 - Ox. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

5.1B - Oxidizing hazardous materials

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service