Skip to Content
Merck

Skip To

55697

5-Hydroxyindole-3-acetic acid

analytical standard

Synonym(s):

5-HIAA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 179,00

About This Item

Empirical Formula (Hill Notation):
C10H9NO3
CAS Number:
Molecular Weight:
191.18
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
168797
MDL number:

€ 179,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

assay

≥98% (HPLC), ≥98% (T)

shelf life

limited shelf life, expiry date on the label

mp

161-164 °C (dec.) (lit.)

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

OC(=O)Cc1c[nH]c2ccc(O)cc12

InChI

1S/C10H9NO3/c12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h1-2,4-5,11-12H,3H2,(H,13,14)

InChI key

DUUGKQCEGZLZNO-UHFFFAOYSA-N

Biochem/physiol Actions

Major serotonin metabolite.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8052968069I3750
format

neat

format

neat

format

neat

format

-

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

-

assay

≥98% (HPLC), ≥98% (T)

assay

≥95.0% (GC), 90.0-105.0% (T)

assay

≥97.5% (GC)

assay

98%

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-


Still not finding the right product?

Explore all of our products under 5-Hydroxyindole-3-acetic acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Shabbir Ahmad et al.
Scientific reports, 10(1), 8925-8925 (2020-06-04)
Melanins are chemically diverse ubiquitous pigments found across the life forms synthesized via different biochemical pathways mainly from L-tyrosine or acetyl CoA. Though few reports suggest the possibility of tryptophan-based melanin synthesis, however, such tryptophan-based melanin and its biosynthesis remained
Ryan T Oglesby et al.
Magnetic resonance in medicine, 84(5), 2389-2399 (2020-04-18)
The diagnosis of monoamine-related psychiatric disorders is based on the phenomenological evaluation of symptoms and behavior by trained clinicians. The CEST technique can be sensitive to monoamines such as serotonin. This study quantifies the CEST properties of the compounds in



Questions

Reviews

No rating value

Active Filters