Skip to Content
Merck

Skip To

68069

Hippuric acid

analytical standard

Synonym(s):

N-Benzoylglycine, Benzoylaminoacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 203,00

About This Item

Linear Formula:
C6H5CONHCH2COOH
CAS Number:
Molecular Weight:
179.17
EC Number:
207-806-3
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
1073987
MDL number:

€ 203,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

mp

187-191 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

OC(=O)CNC(=O)c1ccccc1

InChI

1S/C9H9NO3/c11-8(12)6-10-9(13)7-4-2-1-3-5-7/h1-5H,6H2,(H,10,13)(H,11,12)

InChI key

QIAFMBKCNZACKA-UHFFFAOYSA-N

General description

Hippuric acid, the glycine conjugate of benzoic acid, is formed via a biotransformation process. From scientific literature, it is reported to be found as an excretory product in urine. Hippuric acid is also known to be catabolically synthesized from benzene-type aromatic compounds, believed to be originated from various factors such as environmental exposures, basic human liver metabolism of protein catabolism, cyclic sugar type-compounds, quinic acid, etc.[1]

Application

Hippuric acid may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Human biological fluids using high-performance liquid chromatography (HPLC) with UV detection.
  • Human and rat urine samples using gas chromatography–mass spectrometry (GC–MS).
  • Human urine samples using GC equipped with a flame ionization detector.[2]

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
112003H93808.20649
format

neat

format

-

format

-

format

-

grade

analytical standard

grade

-

grade

-

grade

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

clinical testing

application(s)

peptide synthesis

application(s)

detection

application(s)

-

mp

187-191 °C (lit.)

mp

187-191 °C (lit.)

mp

-

mp

188-190 °C

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Hippuric acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters