Skip to Content
Merck

Skip To

46232

Dipyron monohydrate

VETRANAL®, analytical standard

Synonym(s):

Metamizol, Methapyrone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H16N3NaO4S · H2O
CAS Number:
Molecular Weight:
351.35
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3865381
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical
forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.[Na+].CN(CS([O-])(=O)=O)C1=C(C)N(C)N(C1=O)c2ccccc2

InChI

1S/C13H17N3O4S.Na.H2O/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;;/h4-8H,9H2,1-3H3,(H,18,19,20);;1H2/q;+1;/p-1

InChI key

UNZIDPIPYUMVPA-UHFFFAOYSA-M

Gene Information

General description

Metamizol is an active pharmaceutical ingredient with both analgesic and antipyretic effects, primarily used to relieve pain caused by neuritis and neuralgia.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Metamizol may be used as a reference standard for the determination of metamizol in pharmaceutical formulations by reversed-phase high-performance liquid chromatography (RP-HPLC).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1144PHR179932463
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

analytical standard

format

neat

format

neat

format

neat

format

neat

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

-

Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

100


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Simultaneous Determination Of Metamizole, Thiamin And Pyridoxin In Multicomponent Tablet by RP-HPLC.
Chotimah C.
Journal of Food and Pharmaceutical Sciences, 2(3) (2014)
Natália Amanda Gomes et al.
Archives of oral biology, 122, 105000-105000 (2020-12-12)
This study aimed to compare the impact of testosterone and estrogen replacement therapy on mandibular bone density and bone-related markers. Forty male adult rats were randomly allocated to 4 experimental groups (n = 10/group): Sham (Control); Orchiectomy; Orchiectomy plus testosterone replacement; and
Tina Kirchner et al.
Mediators of inflammation, 2012, 849136-849136 (2012-04-07)
The formation of neutrophil extracellular traps (NETs) depends on the generation of reactive oxygen species (ROS). Previous studies revealed that both NADPH oxidase and myeloperoxidase (MPO) are required for NET release. However, the contribution of various ROS as well as



Global Trade Item Number

SKUGTIN
46232-250MG04061832346427

Questions

Reviews

No rating value

Active Filters