Skip to Content
Merck
All Photos(1)

Key Documents

PHR1799

Supelco

Meloxicam

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Meloxicam, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€176.00

€176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C14H13N3O4S2
CAS Number:
Molecular Weight:
351.40
UNSPSC Code:
41116107
NACRES:
NA.24

€176.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 629
traceable to Ph. Eur. Y0001080
traceable to USP 1379401

API family

meloxicam

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,19H,1-2H3,(H,15,16)/b13-11+

InChI key

DWMREKMVXIFPFM-ACCUITESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Meloxicam is an oxicam derivative and belongs to the group of non-steroidal anti-inflammatory drugs (NSAID), used for its analgesic, anti-inflammatory, and antipyretic properties. It acts by inhibiting the activity of prostaglandin synthetase, thereby preventing the synthesis of prostaglandins and decreasing inflammation.[1]

Application

This pharmaceutical secondary standard can also be used as follows:

  • Development of three spectrophotometric methods to determine meloxicam in its pharmaceutical formulations[1]
  • Voltammetric determination of meloxicam in pharmaceutical formulations using a boron-doped diamond electrode (BDDE)[2]
  • High-performance liquid chromatography-based quantitative analysis of meloxicam in its different tablet formulations[3]
  • Development and validation of a reversed-phase HPLC method for the separation and estimation of meloxicam in its pharmaceutical dosage forms[4]
  • Quantification of meloxicam and piroxicam from plasma samples using a liquid chromatography method combined with UV detection[5]

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7807 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reverse phase HPLC method development and validation for determination of Meloxicam in bulk and pharmaceutical formulation
Shaji J and Jain V
International Journal of Pharmaceutical Sciences and Research, 2, 107-107 (2011)
High-performance liquid chromatography determination of meloxicam and piroxicam with ultraviolet detection
Cox S, et al.
Chromatography Research International, 2014 (2014)
Electrochemical oxidation of anti-inflammatory drug meloxicam and its determination using boron doped diamond electrode
Selesovska R, et al.
Journal of Electroanalytical Chemistry, 858, 113758-113758 (2020)
Spectrophotometric estimation of meloxicam in bulk and its pharmaceutical formulations
Dhandapani B, et al.
International Journal of Pharma Sciences and Research, 1, 217-221 (2010)
Specific and simple hplc assay of ecofriendly meloxicam in pharmaceutical formulations
Mahmood KT, et al.
Journal of Pharmaceutical Sciences and Research, 2, 878-878 (2010)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service